Merck
  • α-Hydroxy-β-keto acid rearrangement-decarboxylation: impact on thiamine diphosphate-dependent enzymatic transformations.
α-Hydroxy-β-keto acid rearrangement-decarboxylation: impact on thiamine diphosphate-dependent enzymatic transformations.
Organic & biomolecular chemistry (2012-11-23)
Maryam Beigi, Sabrina Loschonsky, Patrizia Lehwald, Volker Brecht, Susana L A Andrade, Finian J Leeper, Werner Hummel, Michael Müller
ZUSAMMENFASSUNG
The thiamine diphosphate (ThDP) dependent MenD catalyzes the reaction of α-ketoglutarate with pyruvate to selectively form 4-hydroxy-5-oxohexanoic acid 2, which seems to be inconsistent with the assumed acyl donor role of the physiological substrate α-KG. In contrast the reaction of α-ketoglutarate with acetaldehyde gives exclusively the expected 5-hydroxy-4-oxo regioisomer 1. These reactions were studied by NMR and CD spectroscopy, which revealed that with pyruvate the observed regioselectivity is due to the rearrangement-decarboxylation of the initially formed α-hydroxy-β-keto acid rather than a donor-acceptor substrate role variation. Further experiments with other ThDP-dependent enzymes, YerE, SucA, and CDH, verified that this degenerate decarboxylation can be linked to the reduced enantioselectivity of acyloins often observed in ThDP-dependent enzymatic transformations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiaminpyrophosphat, ≥95%
Sigma-Aldrich
α-Ketoglutarsäure, 99.0-101.0% (T)
Sigma-Aldrich
α-Ketoglutarsäure, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutarsäure Dinatriumsalz Dihydrat, ≥98.0% (dried material, NT)
Sigma-Aldrich
α-Ketoglutarsäure, ≥98.5% (NaOH, titration)
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
Sigma-Aldrich
α-Ketoglutarsäure Natriumsalz, ≥98% (titration)
Sigma-Aldrich
Pyruvate Oxidase from microorganisms, lyophilized powder, ≥1.5 U/mg
Sigma-Aldrich
Pyruvate Oxidase from Aerococcus sp., lyophilized powder, ≥35 units/mg protein (biuret)
Sigma-Aldrich
α-Ketoglutarsäure Natriumsalz, BioUltra
Supelco
α-Ketoglutarsäure Dinatriumsalz Hydrat, ≥95%