Merck
  • Monoamine oxidase A and B inhibiting effect and molecular modeling of some synthesized coumarin derivatives.
Monoamine oxidase A and B inhibiting effect and molecular modeling of some synthesized coumarin derivatives.
Neurochemistry international (2012-11-28)
Omaima M Abdelhafez, Kamelia M Amin, Hamed I Ali, Mohamed M Abdalla, Rasha Z Batran
ZUSAMMENFASSUNG
New series of bioactive 7-oxycoumarin derivatives were synthesized and tested for their in vitro and in vivo monoamine oxidase (MAO) A and B inhibitory effect. In vitro studies revealed exceptionally potent and selective MAO-A inhibitors with K(i) values on a picomolar range. The acetohydrazide (3b) and the dioxopyrrolidine derivative (7b) showed the most potent in vitro and in vivo MAO inhibition activity. Moreover, molecular modeling study of the synthesized compounds into MAO-A (PDB: 2Z5X) and MAO-B (PDB: 2XFN) binding sites exhibited direct correlation between AutoDock binding affinity and% inhibition MAO-A (pM) and MAO-B (μM). In addition, the results of in vivo MAO inhibiting properties (ED(50)) of the tested compounds revealed better direct correlation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Monoaminoxidase A human, recombinant, expressed in baculovirus infected BTI insect cells