- Inhibitory effects of vialinin A and its analog on tumor necrosis factor-α release and production from RBL-2H3 cells.
Inhibitory effects of vialinin A and its analog on tumor necrosis factor-α release and production from RBL-2H3 cells.
Cellular immunology (2012-12-19)
Jun-ichi Onose, Yasukiyo Yoshioka, Yue Qi Ye, Kouichi Sugaya, Arata Yajima, Kayoko Taniguchi, Kiyoshi Okada, Shunsuke Yajima, Shunya Takahashi, Hiroyuki Koshino, Naoki Abe
PMID23246504
ZUSAMMENFASSUNG
Vialinin A is an extremely potent inhibitor of tumor necrosis factor (TNF)-α release from RBL-2H3 cells. The present study investigated in detail the inhibitory effects of vialinin A and its analog, 5',6'-dimethyl-1,1':4',1″-terphenyl-2',3',4,4″-tetraol (DMT), on TNF-α. Vialinin A and DMT inhibited the release of TNF-α from RBL-2H3 cells in a dose-dependent manner, but had no effect on β-hexosaminidase activity. Also, vialinins had little effect on TNF-α mRNA levels. Intriguingly, vialinins inhibited TNF-α production at low concentrations, but not shown a dose-dependency. The potent inhibitory activities of vialinins against TNF-α production and release suggest promising new candidate pathways for anti-inflammatory agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-N-Acetylglucosaminidase aus Canavalia ensiformis (Jack bean), ammonium sulfate suspension, ≥8 units/mg protein
Sigma-Aldrich
β-N-Acetylglucosaminidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution