Merck
Structure of gentlyase, the neutral metalloprotease of Paenibacillus polymyxa.
Acta crystallographica. Section D, Biological crystallography (2013-01-01)
Armin Ruf, Martine Stihle, Jörg Benz, Manfred Schmidt, Harald Sobek
ZUSAMMENFASSUNG
Gentlyase is a bacterial extracellular metalloprotease that is widely applied in cell culture and for tissue dissociation and that belongs to the family of thermolysin-like proteases. The structure of thermolysin has been known since 1972 and that of Bacillus cereus neutral protease since 1992. However, the structure determination of other Bacillus neutral proteases has been hindered by their tendency to cannibalistic autolysis. High calcium conditions that allow the concentration and crystallization of the active Gentlyase metalloprotease without autoproteolysis were identified using thermal fluorescent shift assays. X-ray structures of the protease were solved in the absence and in the presence of the inhibitor phosphoramidon at 1.59 and 1.76 Å resolution, respectively. No domain movement was observed upon inhibitor binding, although such movement is thought to be a general feature of the thermolysin-like protease family. Further analysis of the structure shows that the observed calcium dependency of Gentlyase stability may arise from a partly degenerated calcium site Ca1-2 and a deletion near site Ca3.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, Type X, lyophilized powder, 30-350 units/mg protein (E1%/280)
Sigma-Aldrich
Thermolysin from Geobacillus stearothermophilus, powder, BioReagent, 30-350 units/mg protein (E1%/280), suitable for cell culture