Merck
Transcriptional regulation of fatty acid biosynthesis in Lactococcus lactis.
Journal of bacteriology (2013-01-01)
Tom H Eckhardt, Dorota Skotnicka, Jan Kok, Oscar P Kuipers
ZUSAMMENFASSUNG
Here we study the influence of the putative fatty acid biosynthesis (FAB) regulator FabT (originally called RmaG [Llmg_1788]) on gene transcription in Lactococcus lactis MG1363. A strain with a knockout mutation of the putative regulator was constructed, and its transcriptome was compared to that of the wild-type strain. Almost all FAB genes were significantly upregulated in the knockout. Using electrophoretic mobility shift assays (EMSAs) and DNase I footprinting, the binding motif of the regulator and the binding locations in the genome were characterized. Fatty acid composition analysis revealed that a strain lacking FabT contained significantly more saturated acyl chains in its phospholipids. This observation demonstrates that the vital pathway of FAB in L. lactis is regulated by the repressor FabT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Desoxyribonuklease I, RNase-frei -Lösung aus Rinderpankreas
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free