Merck
  • Benzimidazole analogs of (L)-tryptophan are substrates and inhibitors of tryptophan indole lyase from Escherichia coli.
Benzimidazole analogs of (L)-tryptophan are substrates and inhibitors of tryptophan indole lyase from Escherichia coli.
The FEBS journal (2013-02-27)
Austin P Harris, Robert S Phillips
ZUSAMMENFASSUNG
Tryptophan indole lyase (TIL), an enzyme found in Escherichia coli and related enterobacteria, produces indole from l-tryptophan (l-Trp). Indole is a signaling molecule in bacteria, affecting biofilm formation, pathogenicity and antibiotic resistance. β-(Benzimidazol-1-yl)-l-alanine (BZI-Ala), 2-amino-4-(benzimidazol-1-yl)butyric acid (homo-BZI-Ala) and 2-amino-5-(benzimidazol-1-yl)pentanoic acid (bishomo-BZI-Ala) were synthesized and tested as substrates and inhibitors of TIL. BZI-Ala is a good substrate of TIL, with Km = 300 μm, kcat = 5.6 s(-1) and kcat /Km = 1.9 × 10(4) , similar to l-Trp. BZI-Ala is also a good substrate for H463F mutant TIL, which has very low activity with l-Trp. In contrast, homo-BZI-Ala was found to be a potent competitive inhibitor of TIL, with a Ki of 13.4 μm. However, the higher homolog, bishomo-BZI-Ala, was inactive as an inhibitor of TIL at a concentration of 600 μm, and is thus a much weaker inhibitor. The reaction of TIL with BZI-Ala was too fast to be observed in the stopped-flow spectrophotometer, and shows an aldimine intermediate in the steady state. However, H463F TIL shows equilibrating mixtures of aldimine and quinonoid complexes in the steady state. The spectra of the reaction of TIL with homo-BZI-Ala show a rapidly formed intermediate absorbing at 340 nm, probably a gem-diamine, that decays slowly to form a quinonoid complex absorbing at 494 nm. The potent binding of homo-BZI-Ala may be due to it being a 'bi-product' analog of the indole-α-aminoacrylate complex. These results demonstrate that an amino acid substrate may be converted to a potent inhibitor of TIL simply by homologation, which may be useful in the design of other potent TIL inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Apotryptophanase aus E. coli, soluble powder, 75-150 units/mg solid