Merck
  • Involvement of cyclic AMP receptor protein in regulation of the rmf gene encoding the ribosome modulation factor in Escherichia coli.
Involvement of cyclic AMP receptor protein in regulation of the rmf gene encoding the ribosome modulation factor in Escherichia coli.
Journal of bacteriology (2013-03-12)
Tomohiro Shimada, Hideji Yoshida, Akira Ishihama
ZUSAMMENFASSUNG
The decrease in overall translation in stationary-phase Escherichia coli is accompanied with the formation of functionally inactive 100S ribosomes mediated by the ribosome modulation factor (RMF). At present, however, little is known regarding the regulation of stationary-phase-coupled RMF expression. In the course of a systematic screening of regulation targets of DNA-binding transcription factors from E. coli, we realized that CRP (cyclic AMP [cAMP] receptor protein), the global regulator for carbon source utilization, participates in regulation of some ribosomal protein genes, including the rmf gene. In this study, we carried out detailed analysis of the regulation of the RMF gene by cAMP-CRP. The cAMP-dependent binding of CRP to the rmf gene promoter was confirmed by gel shift and DNase I footprinting assays. By using a reporter assay system, the expression level of RMF was found to decrease in the crp knockout mutant, indicating the involvement of CRP as an activator of the rmf promoter. In good agreement with the reduction of rmf promoter activity, we observed decreases in RMF production and 100S ribosome dimerization in the absence of CRP. Taken together, we propose that CRP regulates transcription activation of the rmf gene for formation of 100S ribosome dimers. Physiological roles of CRP involvement in RMF production are discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Desoxyribonuklease I, RNase-frei -Lösung aus Rinderpankreas
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free