Merck
Probing DNA shape and methylation state on a genomic scale with DNase I.
Proceedings of the National Academy of Sciences of the United States of America (2013-04-12)
Allan Lazarovici, Tianyin Zhou, Anthony Shafer, Ana Carolina Dantas Machado, Todd R Riley, Richard Sandstrom, Peter J Sabo, Yan Lu, Remo Rohs, John A Stamatoyannopoulos, Harmen J Bussemaker
ZUSAMMENFASSUNG
DNA binding proteins find their cognate sequences within genomic DNA through recognition of specific chemical and structural features. Here we demonstrate that high-resolution DNase I cleavage profiles can provide detailed information about the shape and chemical modification status of genomic DNA. Analyzing millions of DNA backbone hydrolysis events on naked genomic DNA, we show that the intrinsic rate of cleavage by DNase I closely tracks the width of the minor groove. Integration of these DNase I cleavage data with bisulfite sequencing data for the same cell type's genome reveals that cleavage directly adjacent to cytosine-phosphate-guanine (CpG) dinucleotides is enhanced at least eightfold by cytosine methylation. This phenomenon we show to be attributable to methylation-induced narrowing of the minor groove. Furthermore, we demonstrate that it enables simultaneous mapping of DNase I hypersensitivity and regional DNA methylation levels using dense in vivo cleavage data. Taken together, our results suggest a general mechanism by which CpG methylation can modulate protein-DNA interaction strength via the remodeling of DNA shape.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II-S, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type II, lyophilized powder, Protein ≥80 %, ≥2,000 units/mg protein
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
Desoxyribonuklease I, RNase-frei -Lösung aus Rinderpankreas
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein
Sigma-Aldrich
Desoxyribonuklease I Rind, recombinant, expressed in Pichia pastoris, lyophilized powder, RNAse and protease, free