Merck
  • Extracellular superoxide dismutase, a potential extracellular biomarker candidate for prolactinoma.
Extracellular superoxide dismutase, a potential extracellular biomarker candidate for prolactinoma.
The West Indian medical journal (2013-04-30)
J Yang, C Zhang, W Zhang, R Shi, Z Zhang
ZUSAMMENFASSUNG
To investigate whether the extracellular superoxide dismutase (EC-SOD) and manganese superoxide dismutase (Mn-SOD) level changes during prolactinoma (PRL) development. Surgical tissues from 37 female patients with PRL were tested for Mn-SOD and serum samples from such PRL patients were tested for EC-SOD level changes with Western Blot. The MnSOD level from blood cells was also investigated to show whether the Mn-SOD variation could locate tumorigenesis tissues. According to the patients' age analysis, age 20-40 years is high risk for getting PRL. There is a positive relationship between the PRL severity and EC-SOD. The Mn-SOD level from surgical tissues, but not blood cells, also shows a corresponding positive relationship to PRL severity, which indicates that elevated Mn-SOD might only happen in PRL tumorigenesis tissues. Extracellular superoxide dismutase is an extracellular protein and the serum EC-SOD could be a good candidate for the diagnoses of prolactinoma.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Sigma-Aldrich
Superoxid-Dismutase Rind, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxid-Dismutase aus Rinderleber, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxid-Dismutase aus Humanerythrocyten, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
Sigma-Aldrich
Superoxid-Dismutase aus Rinderleber, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)