Merck
Cellulose and collagen derived micro-nano structured scaffolds for bone tissue engineering.
Journal of biomedical nanotechnology (2013-04-30)
Aja Aravamudhan, Daisy M Ramos, Jonathan Nip, Matthew D Harmon, Roshan James, Meng Deng, Cato T Laurencin, Xiaojun Yu, Sangamesh G Kumbar
ZUSAMMENFASSUNG
Scaffold based bone tissue engineering (BTE) has made great progress in regenerating lost bone tissue. Materials of natural and synthetic origin have been used for scaffold fabrication. Scaffolds derived from natural polymers offer greater bioactivity and biocompatibility with mammalian tissues to favor tissue healing, due to their similarity to native extracellular matrix (ECM) components. Often it is a challenge to fabricate natural polymer based scaffolds for BTE applications without compromising their bioactivity, while maintaining adequate mechanical properties. In this work, we report the fabrication and characterization of cellulose and collagen based micro-nano structured scaffolds using human osteoblasts (HOB) for BTE applications. These porous micro-nano structured scaffolds (average pore diameter 190 +/- 10 microm) exhibited mechanical properties in the mid range of human trabecular bone (compressive modulus 266.75 +/- 33.22 MPa and strength 12.15 3 +/- 2.23 MPa). These scaffolds supported the greater adhesion and phenotype maintenance of cultured HOB as reflected by higher levels of osteogenic enzyme alkaline phosphatase and mineral deposition compared to control polyester micro-nano structured scaffolds of identical pore properties. These natural polymer based micro-nano structured scaffolds may serve as alternatives to polyester based scaffolds for BTE applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulosepulver, microcrystalline, powder
Sigma-Aldrich
Cellulosepulver, fibers, (medium)
Sigma-Aldrich
Ethylcellulose, 48.0-49.5% (w/w) ethoxyl basis
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Ethylcellulose, 48.0-49.5% (w/w) ethoxyl basis
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Cellulosepulver, colloidal, microcrystalline
Sigma-Aldrich
Ethylcellulose, viscosity 100 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
Ethylcellulose, viscosity 46 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Ethylcellulose, viscosity 22 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Ethylcellulose, viscosity 300 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Ethylcellulose, viscosity 10 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Ethylcellulose, viscosity 4 cP, 5 % in toluene/ethanol 80:20(lit.), extent of labeling: 48% ethoxyl
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Supelco
Cellulosepulver, powder, suitable for column chromatography
Sigma-Aldrich
Phosphatase, alkalisch aus Schweineniere, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Kälberdarmschleimhaut, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)