Merck
  • In vivo antitumor activity of intratumoral fludarabine phosphate in refractory tumors expressing E. coli purine nucleoside phosphorylase.
In vivo antitumor activity of intratumoral fludarabine phosphate in refractory tumors expressing E. coli purine nucleoside phosphorylase.
Cancer chemotherapy and pharmacology (2012-07-05)
Eric J Sorscher, Jeong S Hong, Paula W Allan, William R Waud, William B Parker
ZUSAMMENFASSUNG
Systemically administered fludarabine phosphate (F-araAMP) slows growth of human tumor xenografts that express Escherichia coli purine nucleoside phosphorylase (PNP). However, this treatment has been limited by the amount of F-araAMP that can be administered in vivo. The current study was designed to (1) determine whether efficacy of this overall strategy could be improved by intratumoral administration of F-araAMP, (2) test enhancement of the approach with external beam radiation, and (3) optimize recombinant adenovirus as a means to augment PNP delivery and bystander killing in vivo. The effects of systemic or intratumoral F-araAMP in mice were investigated with human tumor xenografts (300 mg), in which 10 % of the cells expressed E. coli PNP from a lentiviral promoter. Tumors injected with an adenoviral vector expressing E. coli PNP (Ad/PNP; 2 × 10(11) viral particles, 2 times per day × 3 days) and the impact of radiotherapy on tumors treated by this approach were also studied. Radiolabeled F-araAMP was used to monitor prodrug activation in vivo. Intratumoral administration of F-araAMP in human tumor xenografts expressing E. coli PNP resulted in complete regressions and/or prolonged tumor inhibition. External beam radiation significantly augmented this effect. Injection of large human tumor xenografts (human glioma, nonsmall cell lung cancer, or malignant prostate tumors) with Ad/PNP followed by intratumoral F-araAMP resulted in excellent antitumor activity superior to that observed following systemic administration of prodrug. Activation of F-araAMP by E. coli PNP results in destruction of large tumor xenografts in vivo, augments radiotherapy, and promotes robust bystander killing. Our results indicate that intratumoral injection of F-araAMP leads to ablation of tumors in vivo with minimal toxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nukleosidphosphorylase bacterial, recombinant, expressed in E. coli, ≥10 units/mg protein