Merck
Cloning, expression, and purification of a Cu/Zn superoxide dismutase from Jatropha curcas.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2013-05-11)
Ou-Yang Chao, Shun Gao, Feng Cai, Tsair-Wang Chung, Sheng-Hua Wang, Fang Chen
ZUSAMMENFASSUNG
We report cDNA cloning, expression, purification, and characterization of a novel Cu/ Zn superoxide dismutase (SOD) from Jatropha curcas leaves. The full-length cDNA of this SOD contained a 496-bp open-reading frame (ORF) encoding 162 amino acid residues. The recombinant plasmid containing the SOD coding sequence was introduced into Escherichia coli, and the SOD was expressed as a fusion protein. The recombinant SOD was purified from a high-density fed-batch culture using a combination of immobilized metal ion affinity chromatography (IMAC) and Sephadex G25 desalting chromatography. Sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS) analysis indicated that the recombinant SOD was a monomeric protein with a molecular mass of approximately 16.4 kDa. Isoelectric focusing showed that this SOD was a basic protein with pI values of 7.04, 7.33, 8.62, and 8.77. The activity of the SOD was stable at 70 degrees C for 10 min, and in a broad pH range from 4 to 9. The presence of urea (up to 8 M), guanidinium chloride (up to 6 M), and 2-mercaptoethanol (up to 8 mM) had little effect on the activity. The activity decreased gradually with increasing concentrations of imidazole, hydrogen peroxide, and ethylenediaminetetraacetic acid (EDTA). Atomic absorption spectrometry showed the presence of 0.239 copper and 0.258 zinc atoms, respectively, in the SOD polypeptide.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, lyophilized powder, ≥3,000 units/mg protein, Protein ≥95 % by biuret
Sigma-Aldrich
Superoxid-Dismutase Rind, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Sigma-Aldrich
Superoxid-Dismutase aus Rinderleber, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Superoxid-Dismutase aus Humanerythrocyten, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
Sigma-Aldrich
Superoxid-Dismutase aus Rinderleber, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
Sigma-Aldrich
Superoxid-Dismutase aus Rindererythrocyten, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)