- Design, synthesis, and biological evaluation of acetophenone derivatives as dual binding acetylcholinesterase inhibitors.
Design, synthesis, and biological evaluation of acetophenone derivatives as dual binding acetylcholinesterase inhibitors.
Die Pharmazie (2013-06-28)
Yanhong Shen, Baoli Li, Hu Xu, Guoqiang Zhang
PMID23802426
ZUSAMMENFASSUNG
As part of a project aimed at developing new agents for potential application in Alzheimer's disease, a new series of acetophenone derivatives which possess alkylamine side chains were designed, synthesized and assayed as acetylcholinesterase (AChE) inhibitors that could simultaneously bind to the peripheral and catalytic sites of the enzyme. The compounds were synthesized, and the inhibitory activities toward AChE and butyrylcholinesterase (BuChE) in vitro were determined using a modified Ellman method. Of the compounds tested, 6 derivatives were found to inhibit AChE in the micromolar range. The best compound, 2e, had an 1C50 of 0.13 microM. A detailed molecular modeling study was performed to explore the interaction of 2e with AChE.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetylcholinesterase aus Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase aus Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (BCA)
Sigma-Aldrich
Acetylcholinesterase aus Humanerythrocyten, buffered aqueous solution, ≥500 units/mg protein (BCA)