Merck
  • Overexpression of the halophyte Kalidium foliatum H⁺-pyrophosphatase gene confers salt and drought tolerance in Arabidopsis thaliana.
Overexpression of the halophyte Kalidium foliatum H⁺-pyrophosphatase gene confers salt and drought tolerance in Arabidopsis thaliana.
Molecular biology reports (2012-04-28)
Manhong Yao, Youling Zeng, Lin Liu, Yunlan Huang, Enfeng Zhao, Fuchun Zhang
ZUSAMMENFASSUNG
According to sequences of H(+)-pyrophosphatase genes from GenBank, a new H(+)-pyrophosphatase gene (KfVP1) from the halophyte Kalidium foliatum, a very salt-tolerant shrub that is highly succulent, was obtained by using reverse transcription PCR and rapid amplification of cDNA ends methods. The obtained KfVP1 cDNA contained a 2295 bp ORF and a 242 bp 3'-untranslated region. It encoded 764 amino acids with a calculated molecular mass of 79.78 kDa. The deduced amino acid sequence showed high identity to those of H(+)-PPase of some Chenopodiaceae plant species. Semi-quantitative PCR results revealed that transcription of KfVP1 in K. foliatum was induced by NaCl, ABA and PEG stress. Transgenic lines of A. thaliana with 35S::KfVP1 were generated. Three transgenic lines grew more vigorous than the wild type (ecotype Col-0) under salt and drought stress. Moreover, the transgenic plants accumulated more Na(+) in the leaves compared to wild type plants. These results demonstrated that KfVP1 from K. foliatum may be a functional tonoplast H(+)-pyrophosphatase in contributing to salt and drought tolerance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrophosphatase, anorganisch aus Backhefe (S. cerevisiae), powder, ≥500 units/mg protein (E1%/280)
Sigma-Aldrich
Pyrophosphatase, anorganisch aus E. coli, recombinant, expressed in E. coli, lyophilized powder, ≥90%, ≥800 units/mg protein