Merck
  • Crystal structure of human group X secreted phospholipase A2. Electrostatically neutral interfacial surface targets zwitterionic membranes.
Crystal structure of human group X secreted phospholipase A2. Electrostatically neutral interfacial surface targets zwitterionic membranes.
The Journal of biological chemistry (2002-08-06)
Ying H Pan, Bao-Zhu Yu, Alan G Singer, Farideh Ghomashchi, Gerard Lambeau, Michael H Gelb, Mahendra K Jain, Brian J Bahnson
ZUSAMMENFASSUNG
The crystal structure of human group X (hGX) secreted phospholipase A2 (sPLA2) has been solved to a resolution of 1.97 A. As expected the protein fold is similar to previously reported sPLA2 structures. The active site architecture, including the positions of the catalytic residues and the first and second shell water around the Ca2+ cofactor, are highly conserved and remarkably similar to the group IB and group IIA enzymes. Differences are seen in the structures following the (1-12)-N-terminal helix and at the C terminus. These regions are proposed to interact with the substrate membrane surface. The opening to the active site slot is considerably larger in hGX than in human group IIA sPLA2. Furthermore, the electrostatic surface potential of the hGX interfacial-binding surface does not resemble that of the human group IIA sPLA2; the former is highly neutral, whereas the latter is highly cationic. The cationic residues on this face of group IB and IIA enzymes have been implicated in membrane binding and in k(cat*) allostery. In contrast, hGX does not show activation by the anionic charge at the lipid interface when acting on phospholipid vesicles or short-chain phospholipid micelles. Together, the crystal structure and kinetic results of hGX supports the conclusion that it is as active on zwitterionic as on anionic interfaces, and thus it is predicted to target the zwitterionic membrane surfaces of mammalian cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phospholipase A2 aus Honigbienengift (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Phospholipase A2 aus Schweinepankreas, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
Phospholipase A2 aus Rinderpankreas, lyophilized powder, ≥20 units/mg protein