Merck
  • Pattern recognition receptors and interleukin-8 mediate effects of Gram-positive and Gram-negative bacteria on lung epithelial cell function.
Pattern recognition receptors and interleukin-8 mediate effects of Gram-positive and Gram-negative bacteria on lung epithelial cell function.
British journal of pharmacology (2008-06-10)
R Sorrentino, P M de Souza, S Sriskandan, C Duffin, M J Paul-Clark, J A Mitchell
ZUSAMMENFASSUNG
Lung epithelial cells express pattern recognition receptors, which react to bacteria. We have evaluated the effect of Gram-positive and Gram-negative bacteria on interleukin-8 (CXCL8) release from epithelial cells and the integrity of the epithelial barrier. Primary cultures of human airway epithelial cells and the epithelial cell line A549 were used, and CXCL8 release was measured after exposure to Gram-negative or Gram-positive bacteria. Epithelial barrier function was assessed in monolayer cultures of A549 cells. Gram-positive bacteria Staphylococcus aureus or Streptococcus pneumoniae, induced release of CXCL8 from human airway epithelial cells. These bacteria also disrupted barrier function in A549 cells, an effect mimicked by CXCL8 and blocked by specific binding antibodies to CXCL8. Gram-negative bacteria Escherichia coli or Pseudomonas aeruginosa induced greater release of CXCL8 than Gram-positive bacteria. However, Gram-negative bacteria did not affect epithelial barrier function directly, but prevented disruption induced by Gram-positive bacteria. These effects of Gram-negative bacteria on barrier function were mimicked by FK565, an agonist of the nucleotide-binding oligomerization domain 1 (NOD1) receptor, but not by the Toll-like receptor (TLR) 4 agonist bacterial lipopolysaccharide. Neither the Gram-negative bacteria nor FK565 blocked CXCL8 release. These data show differential functional responses induced by Gram-negative and Gram-positive bacteria in human lung epithelial cells. The NOD1 receptors may have a role in preventing disruption of the epithelial barrier in lung, during inflammatory states.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Peroxidase aus Meerrettich, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, lyophilized, powder, ~150 U/mg
Sigma-Aldrich
Peroxidase aus Meerrettich, Type I, essentially salt-free, lyophilized powder, ≥50 units/mg solid (using pyrogallol)
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Peroxidase aus Meerrettich, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Peroxidase aus Meerrettich, Type X, ammonium sulfate suspension
Supelco
Pyrogallol, analytical standard