Merck
  • Activation of human IK and SK Ca2+ -activated K+ channels by NS309 (6,7-dichloro-1H-indole-2,3-dione 3-oxime).
Activation of human IK and SK Ca2+ -activated K+ channels by NS309 (6,7-dichloro-1H-indole-2,3-dione 3-oxime).
Biochimica et biophysica acta (2004-10-09)
Dorte Strøbaek, Lene Teuber, Tino D Jørgensen, Philip K Ahring, Katrine Kjaer, Rie S Hansen, Søren Peter Olesen, Palle Christophersen, Bo Skaaning-Jensen
ZUSAMMENFASSUNG
We have identified and characterized the compound NS309 (6,7-dichloro-1H-indole-2,3-dione 3-oxime) as a potent activator of human Ca2+ -activated K+ channels of SK and IK types, whereas it is devoid of effect on BK type channels. IK- and SK-channels have previously been reported to be activated by the benzimidazolinone, 1-EBIO and more potently by its dichloronated-analogue, DC-EBIO. NS309 is at least 1000 times more potent than 1-EBIO and at least 30 times more potent than DC-EBIO when the compounds are compared on the same cell.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-EBIO, ≥98% (HPLC)