Merck
Synthesis and cytotoxicity study of alkannin derivatives.
European journal of medicinal chemistry (2004-09-01)
Zhi-Shu Huang, Hai-Qiang Wu, Zhi-Fang Duan, Bing-Fen Xie, Zong-Chao Liu, Gong-Kan Feng, Lian-Quan Gu, Albert S C Chan, Yue-Ming Li
ZUSAMMENFASSUNG
Alkannin derivatives (3-19) were prepared through the reaction of beta,beta-dimethylacrylalkannin (1), the most abundant isohexenylnaphthazarin isolated from the roots of Arnebia euchroma, with different types of nucleophiles such as amines and thiols in the absence or presence of a reducing agent. The cytotoxicities of 1-8, 10-14 and 19 against four human carcinoma cell line (GLC-82, CNE2, Bel-7402, K-562) were found to be markedly higher than that of the naturally occurring beta,beta-dimethylacrylalkannin (1) and acetylalkannin (2). This study also shed light on the understanding of the biological activities in terms of the chemical reactivity of alkannins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5,8-Dihydroxy-1,4-naphthochinon, technical grade