Merck
  • Sensitive extractive spectrophotometric methods for the determination of trazodone hydrochloride in pharmaceutical formulations.
Sensitive extractive spectrophotometric methods for the determination of trazodone hydrochloride in pharmaceutical formulations.
Chemical & pharmaceutical bulletin (2006-07-05)
Ramanaboyina Sudhir Kumar, Devagondanahalli Hadagali Manjunatha, Sarfaraj Mohd Takhi Shaikh, Jaldappa Seetharamappa, Kasalanti Harikrishna
ZUSAMMENFASSUNG
Two simple, rapid and sensitive extractive spectrophotometric methods have been developed for the assay of trazodone hydrochloride (TRH) in pure and pharmaceutical formulations. These methods are based on the formation of chloroform soluble ion-association complexes of TRH with bromothymol blue (BTB) and with bromocresol purple (BCP) in KCl-HCl buffer of pH 2.0 (for BTB) and in NaOAc-AcOH buffer of pH of 3.6 (for BCP) with absorption maximum at 423 nm and at 408 nm for BTB and BCP, respectively. Reaction conditions were optimized to obtain the maximum color intensity. The absorbance was found to increase linearly with increase in concentration of TRH, which was corroborated by the calculated correlation coefficient values (0.9996, 0.9945). The systems obeyed Beer's law in the range of 0.2-14.5 and 0.2-14.1 microg/ml for BTB and BCP, respectively. Various analytical parameters have been evaluated and the results have been validated by statistical data. No interference was observed from common excipients present in pharmaceutical formulations. The proposed methods are simple, accurate and suitable for quality control applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bromkresolpurpur, BioReagent, suitable for indicator, Dye content 90 %
Sigma-Aldrich
Bromkresolpurpur, Technical grade