Merck
[Study on quality for Herba Polygoni Orientalis].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2006-10-20)
Ai-min Wang, Yong-lin Wang, Li-na Hu, Yong-jun Li
ZUSAMMENFASSUNG
To establish the qualitative and quantitative detective methods of Herba Polygoni Orientalis. Isorientin, orientin, protocatechuic acid and gallic acid in Herba Polygoni Orientalis were identified by TLC. The contents of isorientin and orientin in Herba Polygoni Orientalis were determined by HPLC. Isorientin, orientin, protocatechuic acid and gallic acid could be identified by TLC. Isorientin and orientin were well separated with Diamonsil C18 column and acetonitrile-0.1% phosphoric acid (18:82) as mobile phase. The linear range of isorientin was 0.075-0.90 microg. The average recovery of isorientin was 98.8% and RSD was 2.1%. The linear range of orientin was 0.041-0.49 microg. The average recovery of orientin was 98.8% and RSD was 2.1%. The methods can be used for qualitative identification and quantitation determination of Herba Polygoni Orientalis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Orientin, ≥97% (HPLC)
Sigma-Aldrich
Isoorientin, ≥98% (HPLC)
Isoorientin, primary reference standard
Orientin, primary reference standard