Merck
Octyl salicylate: a new contact sensitivity.
Contact dermatitis (2006-12-21)
M Singh, M H Beck
ZUSAMMENFASSUNG
We describe acute contact sensitivity to octyl salicylate, an ultraviolet filter, used in many cosmetics and sunscreens.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Octisalat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Ethylhexylsalicylat, 99%
Sigma-Aldrich
2-Ethylhexylsalicylat, ≥99%
Supelco
2-Ethylhexylsalicylat, analytical standard