Merck
Fragrance material review on ethyl hexyl salicylate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
A Lapczynski, D McGinty, L Jones, C S Letizia, A M Api
ZUSAMMENFASSUNG
A toxicologic and dermatologic review of ethyl hexyl salicylate when used as a fragrance ingredient is presented.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Octisalat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Ethylhexylsalicylat, 99%
Sigma-Aldrich
2-Ethylhexylsalicylat, ≥99%
Supelco
2-Ethylhexylsalicylat, analytical standard