Merck
  • Comparative study on two commercial strains of Saccharomyces cerevisiae for optimum ethanol production on industrial scale.
Comparative study on two commercial strains of Saccharomyces cerevisiae for optimum ethanol production on industrial scale.
Journal of biomedicine & biotechnology (2010-05-07)
K Mukhtar, M Asgher, S Afghan, K Hussain, S Zia-Ul-Hussnain
ZUSAMMENFASSUNG
Two commercial strains of Saccharomyces cerevisiae, Saf-Instant (Baker's yeast) and Ethanol red (Mutant) were compared for ethanol production during hot summer season, using molasses diluted up to 6-7 degrees Brix containing 4%-5% sugars. The yeasts were then propagated in fermentation vessels to study the effects of yeast cell count and varying concentrations of Urea, DAP, inoculum size and Lactrol (Antibiotic). Continuous circulation of mash was maintained for 24 hours and after this fermenter was allowed to stay for a period of 16 hours to give time for maximum conversion of sugars into ethanol. Saccharomyces cerevisiae strain (Saf-instant) with cell concentration of 400 millions/mL at molasses sugar level of 13%-15% (pH 4.6 +/- 0.2, Temp. 32 degrees C +/- 1), inoculum size of 25% (v/v), urea concentration, 150 ppm, DAP, 53.4 ppm and Lactrol,150 ppm supported maximum ethanol production (8.8%) with YP/S = 250 L ethanol per tone molasses (96.5% yield), and had significantly lower concentrations of byproducts. By selecting higher ethanol yielding yeast strain and optimizing the fermentation parameters both yield and economics of the fermentation process can be improved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumphosphat, einbasig, ACS reagent, ≥98%
Sigma-Aldrich
Ammoniumphosphat, ACS reagent, ≥98%
Sigma-Aldrich
Ammoniumphosphat, einbasig, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Ammoniumphosphat, reagent grade, ≥98.0%
Sigma-Aldrich
Ammoniumphosphat, einbasig, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumdihydrogenphosphat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniumphosphat, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Ammoniumphosphat, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumphosphat, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammoniumphosphat, einbasig, BioUltra, ≥99.5% (T)
Supelco
Ammoniumphosphat, einbasig, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Ammoniumphosphat, einbasig, suitable for plant cell culture