Merck
  • Anion effect in the diastereoselective formation of bischelated Ni(II) complexes with a novel, chiral phosphine derived from 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU).
Anion effect in the diastereoselective formation of bischelated Ni(II) complexes with a novel, chiral phosphine derived from 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU).
Dalton transactions (Cambridge, England : 2003) (2010-05-08)
Matthew O'Reilly, Roberto Pattacini, Pierre Braunstein
ZUSAMMENFASSUNG
With the new chiral phosphine ligand DBUP, obtained from DBU and Ph(2)PCl, a remarkable anion effect was observed on the diastereoselective formation of the bischelated complexes [Ni(DBUP)(2)](X)(n) (X = Cl, Br, n = 2; X = NiCl(4), n = 1), which are active ethylene oligomerization catalyst precursors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-en (1,5-5), 98%
Sigma-Aldrich
1,8-Diazabicyclo[5.4.0]undec-7-en, puriss., ≥99.0% (GC)