Merck
  • Amine functional monodisperse microbeads via precipitation polymerization of N-vinyl formamide: immobilized laccase for benzidine based dyes degradation.
Amine functional monodisperse microbeads via precipitation polymerization of N-vinyl formamide: immobilized laccase for benzidine based dyes degradation.
Bioresource technology (2011-05-03)
Bunyamin Karagoz, Gulay Bayramoglu, Begum Altintas, Niyazi Bicak, M Yakup Arica
ZUSAMMENFASSUNG
Densely cross-linked poly(vinylamine) microbeads (∼ 2 μm) were prepared by precipitation copolymerization of N-vinyl formamide and ethylene glycoldimethacrylate in acetonitrile. The formamido groups of the microbeads were hydrolyzed into amino groups. Then, amino-functionalized microbeads were used for covalent immobilization of laccase via glutaraldehyde coupling. The average amount of immobilized enzyme was 18.7 mg/g microbeads. Kinetic parameters, V(max) and K(m) values were determined as 20.7 U/mg protein and 2.76 × 10(-2)mmol/L for free enzyme and 15.8 U/mg protein and 4.65 mmol/L for the immobilized laccase, respectively. The immobilized laccase was operated in a batch reactor for the degradation of two different benzidine based dyes (i.e., Direct Blue 1 and Direct Red 128). The laccase immobilized on the microbeads was very effective for removal of these dyes which interfere with the hormonal system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzidin, ≥98%
Sigma-Aldrich
Benzidin -dihydrochlorid, ≥99% (titration)
Supelco
Benzidin, analytical standard
Sigma-Aldrich
Chicago Himmelblau 6B, powder