Merck
Enzyme catalytic promiscuity: The papain-catalyzed Knoevenagel reaction.
Biochimie (2011-10-04)
Wen Hu, Zhi Guan, Xiang Deng, Yan-Hong He
ZUSAMMENFASSUNG
Papain as a sustainable and inexpensive biocatalyst was used for the first time to catalyze the Knoevenagel reactions in DMSO/water. A wide range of aromatic, hetero-aromatic and α,β-unsaturated aldehydes could react with less active methylene compounds acetylacetone and ethyl acetoacetate. The products were obtained in moderate to excellent yields with Z/E selectivities of up to 100:0. This case of biocatalytic promiscuity not only widens the application of papain to new chemical transformations, but also could be developed into a potentially valuable method for organic synthesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetylaceton, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylacetoacetat, ReagentPlus®, 99%
Sigma-Aldrich
Acetylaceton, Wacker Chemie AG, ≥99.5% (GC)
Sigma-Aldrich
Ethylacetoacetat, natural, ≥97%, FG
Sigma-Aldrich
Ethylacetoacetat, puriss. p.a., ≥99.0% (GC)
Supelco
Acetylaceton, analytical standard
Sigma-Aldrich
Ethylacetoacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetoacetat, Arxada quality, ≥99.0% (GC)