Merck
Effectiveness of fresh frozen plasma as supplementary treatment in organophosphate poisoning.
The Medical journal of Malaysia (2012-02-04)
S Pazooki, H Solhi, H R Khoddami Vishteh, S Shadnia, M J Bahram Beigi
ZUSAMMENFASSUNG
With the establishment of the inadequate efficiency of atropines and oximes in reducing morbidity and mortality of patients poisoned by organophosphates, more attention is given to using other methods such as Fresh Frozen Plasma (FFP) as a bioscavenger to mop up organophosphate toxins. This randomized clinical trial was conducted on 56 organophosphate poisoned patients who were randomly assigned to the FFP and control groups in order of admission. The routine treatment in both groups included atropine and, in moderate to severe cases of poisoning, pralidoxime. The FFP group received four packs of FFP as stat dose at the beginning of treatment. No significant difference was seen between the two groups on the atropine and pralidoxime dosage, hospitalization length and mortality. The present study showed that using four packs of FFP as stat dose at the onset of treatment had no significant effect on the clinical course of organophosphate poisoned patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridin-2-Aldoxim-Methochlorid
Sigma-Aldrich
2-Pyridinaldoxim-Methiodid, 99%