Merck
  • Cu(I)-catalyzed annulation for the synthesis of substituted naphthalenes using o-bromobenzaldehydes and β-ketoesters as substrates.
Cu(I)-catalyzed annulation for the synthesis of substituted naphthalenes using o-bromobenzaldehydes and β-ketoesters as substrates.
Organic & biomolecular chemistry (2012-04-14)
Chandi C Malakar, Kavitha Sudheendran, Hans-Georg Imrich, Sabine Mika, Uwe Beifuss
ZUSAMMENFASSUNG
Cu(I)-catalyzed reaction of o-bromobenzaldehydes with β-ketoesters using Cs(2)CO(3) as a base and 2-picolinic acid as an additive proceeds under mild conditions and gives access to substituted naphthalenes in a single step with yields ranging from 71 to 86%. The new annulation process relies on a domino Knoevenagel condensation/C-arylation/1,2-addition/carboxylic acid cleavage. The annulation can also be achieved with o-iodobenzaldehyde.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(I)-Iodid, 98%
Sigma-Aldrich
Benzaldehyd, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyd, purified by redistillation, ≥99.5%
Supelco
Benzaldehyd, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kupfer(I)-Iodid, 99.999% trace metals basis
Sigma-Aldrich
Kupfer(I)-Iodid, purum, ≥99.5%
Sigma-Aldrich
Benzaldehyd, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyd, puriss. p.a., ≥99.0% (GC)
Supelco
Benzaldehyd, analytical standard
Sigma-Aldrich
Benzaldehyd, natural, FCC, FG
Benzaldehyd, European Pharmacopoeia (EP) Reference Standard