Merck
  • Naphthazarin has a protective effect on the 1-methyl-4-phenyl-1,2,3,4-tetrahydropyridine-induced Parkinson's disease model.
Naphthazarin has a protective effect on the 1-methyl-4-phenyl-1,2,3,4-tetrahydropyridine-induced Parkinson's disease model.
Journal of neuroscience research (2012-04-20)
Seon Young Choi, Tae Gen Son, Hee Ra Park, Young Jung Jang, Shin Bi Oh, Bora Jin, Jaewon Lee
ZUSAMMENFASSUNG
"Neurohormesis" refers to a response to a moderate level of stress that enhances the ability of the nervous systems to resist more severe stress that might be lethal or cause dysfunction or disease. Neurohormetic phytochemicals, such as, resveratrol, sulforaphane, curcumin, and catechins, protect neurons against injury and disease. Naphthoquinones, such as, juglone and plumbagin, induce robust hormetic stress responses. However, the possibility that subtoxic dose of 5,8-dihydroxy-1,4-naphthoquinone (naphthazarin) may protect against brain diseases via the activation of an adaptive stress response pathway in the brain has not been investigated. In this study, we examined the neurohormetic effect of a subtoxic dose of naphthazarin in a Parkinson's disease model. It was found that, under these conditions, naphthazarin enhanced movement ability, prevented loss of dopaminergic neurons, and attenuated neuroinflammation in a 1-methyl-4-phenyl-1,2,3,4-tetrahydropyridine-induced Parkinson's disease model. Furthermore, it was found that the neuroprotective effect of naphthazarin was mediated by the suppression of astroglial activation in response to 1-methyl-4-phenylpyridine treatment. In conclusion, we suggest that naphthazarin, in view of its hormetic effect on neuroprotection, be viewed as a potential treatment for Parkinson's disease and other neurodegenerative diseases associated with neuroinflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5,8-Dihydroxy-1,4-naphthochinon, technical grade