Merck
  • Probenecid: novel use as a non-injurious positive inotrope acting via cardiac TRPV2 stimulation.
Probenecid: novel use as a non-injurious positive inotrope acting via cardiac TRPV2 stimulation.
Journal of molecular and cellular cardiology (2012-05-09)
Sheryl E Koch, Xiaoqian Gao, Lauren Haar, Min Jiang, Valerie M Lasko, Nathan Robbins, Wenfeng Cai, Cole Brokamp, Priyanka Varma, Michael Tranter, Yong Liu, Xiaoping Ren, John N Lorenz, Hong-Sheng Wang, W Keith Jones, Jack Rubinstein
ZUSAMMENFASSUNG
Probenecid is a highly lipid soluble benzoic acid derivative originally used to increase serum antibiotic concentrations. It was later discovered to have uricosuric effects and was FDA approved for gout therapy. It has recently been found to be a potent agonist of transient receptor potential vanilloid 2 (TRPV2). We have shown that this receptor is in the cardiomyocyte and report a positive inotropic effect of the drug. Using echocardiography, Langendorff and isolated myocytes, we measured the change in contractility and, using TRPV2(-/-) mice, proved that the effect was mediated by TRPV2 channels in the cardiomyocytes. Analysis of the expression of Ca(2+) handling and β-adrenergic signaling pathway proteins showed that the contractility was not increased through activation of the β-ADR. We propose that the response to probenecid is due to activation of TRPV2 channels secondary to SR release of Ca(2+).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Probenecid
Probenecid, European Pharmacopoeia (EP) Reference Standard