Merck
  • Amphiphilic cholic-acid-modified dextran sulfate and its application for the controlled delivery of superoxide dismutase.
Amphiphilic cholic-acid-modified dextran sulfate and its application for the controlled delivery of superoxide dismutase.
Macromolecular bioscience (2012-05-19)
Yubing Xiong, Jianing Qi, Ping Yao
ZUSAMMENFASSUNG
A novel amphiphilic and biodegradable polyelectrolyte DS-CA is prepared by the esterification of DS with CA. DS-CA can self-assemble into stable nanoparticles in water. SOD can effectively associate with DS-CA at pH = 5.0 by virtue of electrostatic and hydrophobic interactions. SOD release from the complex nanoparticles is slow at pH = 1.2. The release at pH = 7.4 PBS shows an extended behavior and is tunable by changing the weight ratio of SOD to DS-CA as well as the CA substitution degree. Increasing the CA substitution degree of DS-CA can significantly enhance the cellular uptake of the loaded SOD. This study demonstrates that the amphiphilic DS-CA provides a promising strategy for oral delivery of protein/peptide drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholinsäure, from bovine and/or ovine, ≥98%
Cholinsäure, European Pharmacopoeia (EP) Reference Standard