Merck
  • Chronic Δ⁹-tetrahydrocannabinol exposure induces a sensitization of dopamine D₂/₃ receptors in the mesoaccumbens and nigrostriatal systems.
Chronic Δ⁹-tetrahydrocannabinol exposure induces a sensitization of dopamine D₂/₃ receptors in the mesoaccumbens and nigrostriatal systems.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2012-06-14)
Nathalie Ginovart, Benjamin B Tournier, Marcelle Moulin-Sallanon, Thierry Steimer, Vicente Ibanez, Philippe Millet
ZUSAMMENFASSUNG
Δ⁹-tetrahydrocannabinol (THC), through its action on cannabinoid type-1 receptor (CB₁R), is known to activate dopamine (DA) neurotransmission. Functional evidence of a direct antagonistic interaction between CB₁R and DA D₂-receptors (D₂R) suggests that D₂R may be an important target for the modulation of DA neurotransmission by THC. The current study evaluated, in rodents, the effects of chronic exposure to THC (1 mg/kg/day; 21 days) on D₂R and D₃R availabilities using the D₂R-prefering antagonist and the D₃R-preferring agonist radiotracers [¹⁸F]fallypride and [³H]-(+)-PHNO, respectively. At 24 h after the last THC dose, D₂R and D₃R densities were significantly increased in midbrain. In caudate/putamen (CPu), THC exposure was associated with increased densities of D₂R with no change in D₂R mRNA expression, whereas in nucleus accumbens (NAcc) both D₃R binding and mRNA levels were upregulated. These receptor changes, which were completely reversed in CPu but only partially reversed in NAcc and midbrain at 1 week after THC cessation, correlated with an increased functionality of D₂/₃R in vivo, based on findings of increased locomotor suppressive effect of a presynaptic dose and enhanced locomotor activation produced by a postsynaptic dose of quinpirole. Concomitantly, the observations of a decreased gene expression of tyrosine hydroxylase in midbrain together with a blunted psychomotor response to amphetamine concurred to indicate a diminished presynaptic DA function following THC. These findings indicate that the early period following THC treatment cessation is associated with altered presynaptic D₂/₃R controlling DA synthesis and release in midbrain, with the concurrent development of postsynaptic D₂/₃R supersensitivity in NAcc and CPu. Such D₂/₃R neuroadaptations may contribute to the reinforcing and habit-forming properties of THC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Supelco
Phyllochinon (K1), analytical standard
Supelco
Phytonadion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Quinpirole hydrochloride, ≥98% (HPLC), solid
Phytomenadion, European Pharmacopoeia (EP) Reference Standard