Merck
  • Formation of the two novel glycolipid biosurfactants, mannosylribitol lipid and mannosylarabitol lipid, by Pseudozyma parantarctica JCM 11752T.
Formation of the two novel glycolipid biosurfactants, mannosylribitol lipid and mannosylarabitol lipid, by Pseudozyma parantarctica JCM 11752T.
Applied microbiology and biotechnology (2012-06-23)
Tomotake Morita, Tokuma Fukuoka, Tomohiro Imura, Dai Kitamoto
ZUSAMMENFASSUNG
In order to develop novel glycolipid biosurfactants, Pseudozyma parantarctica JCM 11752(T), which is known as a producer of mannosylerythritol lipids (MEL), was cultivated using different sugar alcohols with the presence of vegetable oil. When cultivated in a medium containing 4 % (w/v) olive oil and 4 % D-ribitol or D-arabitol, the yeast strain provided different glycolipids, compared to the case of no sugar alcohol. On TLC, both of the extracted glycolipid fractions gave two major spots corresponding to MEL-A (di-acetylated MEL) and MEL-B (mono-acetylated MEL). Based on (1)H NMR analysis, one glycolipid was identified as MEL-A, but the other was not MEL-B. On high-performance liquid chromatography after acid hydrolysis, the unknown glycolipid from the D-ribitol culture provided mainly two peaks identical to D-mannose and D-ribitol, and the other unknown glycolipid from the D-arabitol culture did two peaks identical to D-mannose and D-arabitol. Accordingly, the two unknown glycolipids were identified as mannosylribitol lipid (MRL) and mannosylarabitol lipid (MAL), respectively. The observed critical micelle concentration (CMC) and surface tension at CMC of MRL were 1.6 × 10(-6) M and 23.7 mN/m, and those of MAL were 1.5 × 10(-6) M and 24.2 mN/m, respectively. These surface-tension-lowering activities were significantly higher compared to conventional MEL. Furthermore, on a water-penetration scan, MRL and MAL efficiently formed not only the lamella phase (L(α)) but also the myelins at a wide range of concentrations, indicating their excellent self-assembling properties and high hydrophilicity. The present two glycolipids should thus facilitate the application of biosurfactants as new functional materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adonitol, ≥99%
Sigma-Aldrich
L-(−)-Arabit, ≥98% (GC)
Sigma-Aldrich
D-(+)-Arabit, ≥99% (GC)
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
L-Arabinitol, European Pharmacopoeia (EP) Reference Standard