Merck
  • Acute effects of prostaglandin E1 and E2 on vascular reactivity and blood flow in situ in the chick chorioallantoic membrane.
Acute effects of prostaglandin E1 and E2 on vascular reactivity and blood flow in situ in the chick chorioallantoic membrane.
Prostaglandins, leukotrienes, and essential fatty acids (2012-08-04)
D R Harland, L D Lorenz, K Fay, B E Dunn, S K Gruenloh, J Narayanan, E R Jacobs, M Medhora
ZUSAMMENFASSUNG
The chick chorioallantoic membrane (CAM) subserves gas exchange in the developing embryo and shell-less culture affords a unique opportunity for direct observations over time of individual blood vessels to pharmacologic interventions. We tested a number of lipids including prostaglandins PGE(1&2) for vascular effects and signaling in the CAM. Application of PGE(1&2) induced a decrease in the diameter of large blood vessels and a concentration-dependent, localized, reversible loss of blood flow through small vessels. The loss of flow was also mimicked by misoprostol, an agonist for 3 of 4 known PGE receptors, EP(2-4), and by U46619, a thromboxane mimetic. Selective receptor antagonists for EP(3) and thromboxane each partially blocked the response. This is a first report of the effects of prostaglandins on vasoreactivity in the CAM. Our model allows the unique ability to examine simultaneous responses of large and small vessels in real time and in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
9,11-Dideoxy-11α,9α-Epoxymethanoprostaglandin F -Lösung, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid
Sigma-Aldrich
SC 19220, ≥98% (HPLC), solid