- Liquid-liquid interface motion of a capsule motor powered by the interlayer Marangoni effect.
Liquid-liquid interface motion of a capsule motor powered by the interlayer Marangoni effect.
The journal of physical chemistry. B (2012-08-17)
Guanjia Zhao, Martin Pumera
PMID22894756
ZUSAMMENFASSUNG
A novel thin capsule motor has been described in this report. It utilizes the Marangoni effect for the solid capsule to run at a water-oil interlayer, which has not been reported previously. Intrinsic and environment factors influencing the motion were investigated. It is also possible for the velocity, direction, and start/stop of the motion of the capsule to be manipulated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polysulfon, average Mw ~35,000 by LS, average Mn ~16,000 by MO, pellets (Transparent)