Merck
The effect of camphor and borneol on rat thymocyte viability and oxidative stress.
Molecules (Basel, Switzerland) (2012-08-29)
Emiliya Cherneva, Voja Pavlovic, Andrija Smelcerovic, Denitsa Yancheva
ZUSAMMENFASSUNG
Camphor and borneol are wildly distributed in the essential oils of medicinal plants from various parts of the World. Our study has been carried out to evaluate the effect of these two bicyclic monoterpenes on rat thymocytes. Camphor and borneol at concentrations of 0.5 and 5 µg/mL did not induce significant toxicity on the immune system cells, while a significant increase of thymocyte viability was detected when cells were incubated with 50 µg/mL of camphor. A significant increase of cell viability was similarly detected when thymocytes were cultivated with borneol at concentrations of 0.5 and 5 µg/mL. The role of camphor and borneol in reactive oxygen species (ROS) production and mitochondrial membrane potential (MMP) disturbances in rat thymocytes as well as their potential mechanism(s) of action were also discussed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Campher, 96%
Sigma-Aldrich
Isoborneol, 95%
Sigma-Aldrich
(+)-Borneol, 97%
Sigma-Aldrich
(1R)-(+)-Kampfer, 98%
Supelco
D-Campher, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(−)-Borneol, predominantly endo, 97%
Sigma-Aldrich
(1S)-(−)-Kampfer, 95%
Sigma-Aldrich
D-Campher, ≥97%, FG
Supelco
(−)-Borneol, analytical standard
Campher (dl), primary reference standard
Sigma-Aldrich
(±)-Campher, purum, synthetic, ≥95.0% (GC)
Sigma-Aldrich
(±)-Campher, ≥95.5%
Sigma-Aldrich
(±)-Campher, meets analytical specification of Ph. Eur., BP, ≥95% (GC), racemic
Supelco
(−)-Campher, analytical standard
Supelco
(+)-Borneol, analytical standard
Kampfer (racemisch), European Pharmacopoeia (EP) Reference Standard