Merck
  • Acetalization allows the photoheterolysis of the Ar-Cl bond in chlorobenzaldehydes and chloroacetophenones.
Acetalization allows the photoheterolysis of the Ar-Cl bond in chlorobenzaldehydes and chloroacetophenones.
The Journal of organic chemistry (2012-09-12)
Carlotta Raviola, Stefano Protti, Davide Ravelli, Mariella Mella, Angelo Albini, Maurizio Fagnoni
ZUSAMMENFASSUNG
The nonaccessibility of phenyl cations by irradiation of electron-poor aryl chlorides was circumvented by transforming the carbonyl group of aromatic ketones or aldehydes into the corresponding 1,3-dioxolanes and the carboxyl group of benzoate ester into an orthoester functionality. This transformation allowed the heterolytic photoactivation of the Ar-Cl bond in protic media and the generation of phenyl cations. In the presence of π-bond nucleophiles, arylated products were obtained in good to excellent yields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Chlorbenzaldehyd, 97%
Sigma-Aldrich
2-Chlorbenzaldehyd, 99%
Sigma-Aldrich
2-Chlor-acetophenon, 98%
Sigma-Aldrich
2-Chlor-acetophenon, purum, ≥98.0% (GC)