Merck
  • Multinuclear palladium(II) complexes from P-C and P-Cl bond cleavage in (R(P),R(P)/S(P),S(P))-[PdCl2{1,2-(P(t)BuCl)2C2B10H10}].
Multinuclear palladium(II) complexes from P-C and P-Cl bond cleavage in (R(P),R(P)/S(P),S(P))-[PdCl2{1,2-(P(t)BuCl)2C2B10H10}].
Chemical communications (Cambridge, England) (2012-09-13)
Ilham Maulana, Peter Lönnecke, Evamarie Hey-Hawkins
ZUSAMMENFASSUNG
Two multinuclear palladium(II) complexes have been isolated as decomposition products resulting from P-Cl and P-C bond cleavage in (R(P),R(P)/S(P),S(P))-[PdCl(2){1,2-(P(t)BuCl)(2)C(2)B(10)H(10)}] in dichloromethane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichlormethan, analytical standard
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichlormethan, Selectophore, ≥99.5%