Merck
  • An improved high-throughput Nile red fluorescence assay for estimating intracellular lipids in a variety of yeast species.
An improved high-throughput Nile red fluorescence assay for estimating intracellular lipids in a variety of yeast species.
Journal of microbiological methods (2012-09-19)
I R Sitepu, L Ignatia, A K Franz, D M Wong, S A Faulina, M Tsui, A Kanti, K Boundy-Mills
ZUSAMMENFASSUNG
A rapid and inexpensive method for estimating lipid content of yeasts is needed for screening large numbers of yeasts samples. Nile red is a fluorescent lipophilic dye used for detection and quantification of intracellular lipid droplets in various biological system including algae, yeasts and filamentous fungi. However, a published assay for yeast is affected by variable diffusion across the cell membrane, and variation in the time required to reach maximal fluorescence emission. In this study, parameters that may influence the emission were varied to determine optimal assay conditions. An improved assay with a high-throughput capability was developed that includes the addition of dimethyl sulfoxide (DMSO) solvent to improve cell permeability, elimination of the washing step, the reduction of Nile red concentration, kinetic readings rather than single time-point reading, and utilization of a black 96-well microplate. The improved method was validated by comparison to gravimetric determination of lipid content of a broad variety of ascomycete and basidiomycete yeast species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nilrot, suitable for microscopy
Sigma-Aldrich
Nilrot, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Nilrot, Technical grade