Merck
Real-time monitoring of tyrosine hydroxylase activity using a plate reader assay.
Analytical biochemistry (2012-09-27)
Lydia M Vermeer, Colin A Higgins, David L Roman, Jonathan A Doorn
ZUSAMMENFASSUNG
Tyrosine hydroxylase (TH) is the rate-limiting step in dopamine (DA) synthesis, oxidizing tyrosine to l-DOPA, which is further metabolized to DA. Current assays for monitoring activity of this enzyme require extensive work-up, require long analysis time, and measure end points, thereby lacking real-time kinetics. This work presents the development of the first real-time colorimetric assay for determining the activity of TH using a plate reader. The production of l-DOPA is followed using sodium periodate to oxidize l-DOPA to the chromophore dopachrome, which can be monitored at 475 nm. Advantages to this method include decreased sample analysis time, shorter assay work-up, and the ability to run a large number of samples at one time. Furthermore, the assay was adapted for high-throughput screening and demonstrated an excellent Z-factor (> 0.8), indicating suitability of this assay for high-throughput analysis. Overall, this novel assay reduces analysis time, increases sample number, and allows for the study of activity using real-time kinetics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumperiodat, ACS reagent, ≥99.8%
Sigma-Aldrich
Natrium-(Meta-)Periodat, ≥99.0%
Sigma-Aldrich
Periodsäure, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Periodsäure, ACS reagent, 99%
Sigma-Aldrich
Natrium-(Meta-)Periodat, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Periodsäure, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Periodsäure, 99.999% trace metals basis