Merck
  • Microwave assisted synthesis of 2,2'-arylene-substituted bis(4H-3,1-benzoxazin-4-one) derivatives using the complex cyanuric chloride/N,N-dimethylformamide.
Microwave assisted synthesis of 2,2'-arylene-substituted bis(4H-3,1-benzoxazin-4-one) derivatives using the complex cyanuric chloride/N,N-dimethylformamide.
Molecules (Basel, Switzerland) (2012-10-02)
Mehdi Shariat, Mohd Wahid Samsudin, Zuriati Zakaria
ZUSAMMENFASSUNG
A new and efficient method has been designed to prepare 2,2'-arylene-substituted bis(4H-3,1-benzoxazin-4-one) derivatives by using the mixture of cyanuric chloride and N,N-dimethylformamide in a microwave-assisted reaction. The method used and presented here has good rate enhancement and excellent yields.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
Cyanurchlorid, 99%
Sigma-Aldrich
Anthranilsäure, reagent grade, ≥98%
Supelco
N,N-Dimethylformamid, analytical standard
Sigma-Aldrich
Anthranilsäure, puriss. p.a., ≥99.5% (T)