Merck
  • Effects of lauric acid on ruminal protozoal numbers and fermentation pattern and milk production in lactating dairy cows.
Effects of lauric acid on ruminal protozoal numbers and fermentation pattern and milk production in lactating dairy cows.
Journal of animal science (2012-10-26)
A P Faciola, G A Broderick, A Hristov, M I Leão
ZUSAMMENFASSUNG
The objectives of this study were to evaluate lauric acid (LA) as a practical ruminal protozoa-suppressing agent and assess effects of protozoal suppression on fermentation patterns and milk production in dairy cows. In a pilot study, 6 lactating Holstein cows fitted with ruminal cannulae were used in a randomized complete-block design trial. Cows were fed a basal total mixed ration (TMR) containing (DM basis) 15% alfalfa silage, 40% corn silage, 30% rolled high moisture shelled corn, and 14% solvent soybean meal, and assigned to 1 of 3 treatments: 1) control, 2) 160 g/d of LA, or 3) 222 g/d of sodium laurate, which is equimolar to 160 g/d of LA, all given as a single dose into the rumen via cannulae before feeding. Both agents showed high antiprotozoal activity when pulse dosed at these amounts via ruminal cannulae, reducing protozoa by 90% (P<0.01) within 2 d of treatment. Lauric acid reduced ruminal ammonia concentration by 60% (P<0.01) without altering DMI. Both agents reduced ruminal total free AA concentration (P<0.01) and LA did not affect ruminal pH or total VFA concentration. In a large follow-up feeding trial, 52 Holstein cows (8 with ruminal cannulae) were used in a randomized complete-block design trial. Cows were assigned to 1 of 4 diets and fed only that diet throughout the study. The TMR contained (DM basis) 29% alfalfa silage, 36% corn silage, 14% rolled high moisture shelled corn, and 8% solvent soybean meal. The 4 experimental diets were similar, except part of the finely ground dry corn was replaced with LA in stepwise increments from 0 to 0.97% of dietary DM, which provided (as consumed) 0, 83, 164, and 243 g/d of LA. Adding these amounts of LA to the TMR did not affect DMI, ruminal pH, or other ruminal traits, and milk production. However, LA consumed at 164 and 243 g/d in the TMR reduced the protozoal population by only 25% and 30% (P=0.05), respectively, showing that these levels, when added to the TMR, were not sufficient to achieve a concentration within the rumen that promoted the antiprotozoal effect of LA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Laurinsäure, 98%
Sigma-Aldrich
Natrium-dodecanoat, 99-100%
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecansäure, ≥99% (GC/titration)
Supelco
Dodecansäure, analytical standard