Merck
  • Structural basis of the binding of fatty acids to peptidoglycan recognition protein, PGRP-S through second binding site.
Structural basis of the binding of fatty acids to peptidoglycan recognition protein, PGRP-S through second binding site.
Archives of biochemistry and biophysics (2012-11-15)
Pradeep Sharma, Shavait Yamini, Divya Dube, Amar Singh, Gorakh Mal, Nisha Pandey, Mau Sinha, Abhay Kumar Singh, Sharmistha Dey, Punit Kaur, Dipendra K Mitra, Sujata Sharma, Tej P Singh
ZUSAMMENFASSUNG
Short peptidoglycan recognition protein (PGRP-S) is a member of the mammalian innate immune system. PGRP-S from Camelus dromedarius (CPGRP-S) has been shown to bind to lipopolysaccharide (LPS), lipoteichoic acid (LTA) and peptidoglycan (PGN). Its structure consists of four molecules A, B, C and D with ligand binding clefts situated at A-B and C-D contacts. It has been shown that LPS, LTA and PGN bind to CPGRP-S at C-D contact. The cleft at the A-B contact indicated features that suggested a possible binding of fatty acids including mycolic acid of Mycobacterium tuberculosis. Therefore, binding studies of CPGRP-S were carried out with fatty acids, butyric acid, lauric acid, myristic acid, stearic acid and mycolic acid which showed affinities in the range of 10(-5) to 10(-8) M. Structure determinations of the complexes of CPGRP-S with above fatty acids showed that they bound to CPGRP-S in the cleft at the A-B contact. The flow cytometric studies showed that mycolic acid induced the production of pro-inflammatory cytokines, TNF-α and IFN-γ by CD3+ T cells. The concentrations of cytokines increased considerably with increasing concentrations of mycolic acid. However, their levels decreased substantially on adding CPGRP-S.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Buttersäure, ≥99%
Sigma-Aldrich
Laurinsäure, ≥98%, FCC, FG
Sigma-Aldrich
Myristinsäure, Sigma Grade, ≥99%
Sigma-Aldrich
Lipoteichoesäure aus Staphylococcus aureus, bacterial cell wall polymer
Sigma-Aldrich
Laurinsäure, 98%
Supelco
Buttersäure, analytical standard
Sigma-Aldrich
Natrium-dodecanoat, 99-100%
Sigma-Aldrich
Buttersäure, ≥99%, FG
Sigma-Aldrich
Buttersäure, natural, ≥99%, FCC, FG
Sigma-Aldrich
Laurinsäure, natural, ≥98%, FCC, FG
Sigma-Aldrich
Dodecansäure, ≥99% (GC/titration)
Sigma-Aldrich
Myristinsäure, natural, ≥98.5%, FG
Sigma-Aldrich
Lipoteichoic acid from Bacillus subtilis
Sigma-Aldrich
Myristinsäure, ≥95%, FCC, FG
Sigma-Aldrich
Myristinsäure, ≥98.0% (GC)
Sigma-Aldrich
Lipoteichoic acid from Streptococcus pyogenes
Supelco
Myristinsäure, analytical standard
Supelco
Dodecansäure, analytical standard
Supelco
Myristinsäure, Pharmaceutical Secondary Standard; Certified Reference Material