Merck
Camptocormia due to inclusion body myositis.
Journal of clinical neuromuscular disease (2012-11-23)
Brent P Goodman, Teerin Liewluck, Brian A Crum, Robert J Spinner
ZUSAMMENFASSUNG
Inclusion body myositis is the most common idiopathic inflammatory myopathy in elderly individuals. It typically causes proximal and distal limb weakness with forearm flexors and quadriceps being the most severely affected muscles. Axial musculature is infrequently involved. Here, we report an 80-year-old man who presented with an 18-month history of progressive truncal weakness causing stooped posture while standing and walking. Neurologic examination revealed no limb weakness. magnetic resonance imaging studies showed atrophy and findings, suggesting fatty replacement of paraspinal muscles. Needle electromyography confirmed the presence of an axial myopathy. Thoracic paraspinal muscle biopsy showed canonical features of inclusion body myositis. The current patient broadens the clinical presentation of inclusion body myositis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Eosin Y -Lösung, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y Dinatriumsalz, certified by the BSC
Sigma-Aldrich
Eosin Y Dinatriumsalz, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y, 75% (HPLC)