Merck
Studies on the interaction mechanism of pyrene derivatives with human tumor-related DNA.
Molecules (Basel, Switzerland) (2012-11-30)
Li Li, Jia Lu, Chongzheng Xu, Huihui Li, Xiaodi Yang
ZUSAMMENFASSUNG
Pyrene derivatives can be carcinogenic, teratogenic and mutagenic, thus having the potential to cause malignant diseases. In this work, the interactions of two selected pyrene derivatives (1-OHP and 1-PBO) and human tumor-related DNA (p53 DNA and C-myc DNA) are investigated by spectroscopic and non-native polyacrylamide gel electrophoresis (PAGE) methods. Using fluorescence spectrometry and circular dichroism (CD), DNA interactions of pyrene derivatives are confirmed to occur mainly via the groove binding mode supported by the intercalation into the base pairs of DNA. There is an obvious binding order of pyrene derivatives to the targeted DNA, 1-OHP > 1-PBO. The binding constants of 1-OHP are 1.16 × 10(6) L × mol(-1) and 4.04 × 10(5) L × mol(-1) for p53 DNA and C-myc DNA, respectively, while that of 1-PBO are only 2.04 × 10(3) L × mol(-1) and 1.39 × 10(3) L × mol(-1) for p53 DNA and C-myc DNA, respectively. Besides, the binding of pyrene derivatives to p53 DNA is stronger than that for C-myc DNA. CD and PAGE results indicate that the binding of pyrene derivatives can affect the helical structures of DNA and further induce the formation of double-chain antiparallel G-quadruplex DNA of hybrid G-rich sequences.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyren, 98%
Sigma-Aldrich
Pyren, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Supelco
Pyren, analytical standard
Supelco
Pyren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Pyren, sublimed grade, 99%
Pyren, BCR®, certified reference material