Merck
Nano-scale adhesion in multilayered drug eluting stents.
Journal of the mechanical behavior of biomedical materials (2012-12-15)
Sina Youssefian, Nima Rahbar
ZUSAMMENFASSUNG
Using stainless steel 316L for drug-eluting stents needs specific surface finishing due to corrosion phenomena that take place on the metal surface upon prolonged contact with human tissue. Poly (o-chloro-p-xylylene) (Parylene C) is one of the inert and biocompatible materials that are used for 316L coating with γ-methacryloxypropyltrimethoxysilane as an adhesion promoter. In this study, a combination of atomic force microscopy experiments and contact theories have been used to quantify the work of adhesion between parylene C/316L and silane added parylene C/316L. An atomistic simulation has been used, first, to investigate and compare the adhesion at the room temperature with the experiments and then, to investigate the effect of aqueous environment with higher temperature, inside the body, on the adhesion between layers in the structure of drug eluting stent. The simulation results of simplified model for 316L are in good agreement with the experimental results and suggest that the week affiliation between this polymer and 316L is mainly due to Van der Waals interactions. The effect of temperature on the adhesion is found to be regressive and as the water molecules permeate the polymer the adhesion decreases. They also imply that the effect of silane on the adhesion between parylene C and steel is modest.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-(Trimethoxysilyl)propylmethacrylat, 98%
Sigma-Aldrich
3-(Trimethoxysilyl)propylmethacrylat, ≥97%
Sigma-Aldrich
Chrom(III)-oxid, powder, ≥98%
Sigma-Aldrich
Chrom(III)-oxid, powder, 99.9% trace metals basis
Sigma-Aldrich
Chrom(III)-oxid, nanopowder, <100 nm particle size (TEM), 98% trace metals basis