- Magnesium stearate: an underestimated allergen.
Magnesium stearate: an underestimated allergen.
Journal of biological regulators and homeostatic agents (2012-12-18)
A Tammaro, C Abruzzese, A Narcisi, G Cortesi, F Persechino, F R Parisella, S Persechino
PMID23241129
ZUSAMMENFASSUNG
Magnesium stearate is a substance often used as a diluent in the manufacture of medical tablets, capsules and powders. Moreover it is usually found as a food additive or pharmaceutical excipient. We report the first case of a 28 years old woman affected by an allergic reaction from this substance with an urticarial manifestation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Stearinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland