Merck
Effects of succinate and pH on cooked beef color.
Meat science (2013-01-15)
R Ramanathan, R A Mancini, G A Dady, C B Van Buiten
ZUSAMMENFASSUNG
Two experiments were conducted to assess the effects of succinate and pH on cooked beef color. In experiment 1, ten strip loins (M. longissimus lumborum) were divided in half and assigned to either non-enhanced control or 2.5% succinate. Each half-loin was cut into steaks, packaged in vacuum or 80% oxygen, and stored at 1°C for 0, 6, or 12 days. Steaks were cooked to either 66°C or 71°C. Succinate increased (P<0.05) steak pH, raw a* values, and interior cooked redness when packaged in high oxygen. In experiment 2, to assess the role of succinate in raw and cooked color, succinate or ammonium hydroxide was added to ground beef patties to result in a common meat pH (5.9). At a similar pH, succinate had greater metmyoglobin reducing activity and internal cooked redness compared with ammonium hydroxide (P<0.05). In addition to ingredient-based changes in muscle pH, succinate may influence color by regenerating reducing equivalents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniumhydroxid -Lösung, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Ammoniumhydroxid -Lösung, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammoniumhydroxid -Lösung, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)
Supelco
Bernsteinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland