Merck
  • The influence of the galenic form on pharmacokinetics of florfenicol after intramuscular administration in pigs.
The influence of the galenic form on pharmacokinetics of florfenicol after intramuscular administration in pigs.
Journal of veterinary pharmacology and therapeutics (2013-01-18)
J Embrechts, L Sedlák, I Hlavizna, P Heřmanský, I Ćechová, M Brožková, A Beránková, J De Busser
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiamphenicol