Merck
Fatty acid solubilizer from the oral disk of the blowfly.
PloS one (2013-01-18)
Yuko Ishida, Jun Ishibashi, Walter S Leal
ZUSAMMENFASSUNG
Blowflies are economic pests of the wool industry and potential vectors for epidemics. The establishment of a pesticide-free, environmentally friendly blowfly control strategy is necessary. Blowflies must feed on meat in order to initiate the cascade of events that are involved in reproduction including juvenile hormone synthesis, vitellogenesis, and mating. During feeding blowflies regurgitate salivary lipase, which may play a role in releasing fatty acids from triglycerides that are found in food. However, long-chain fatty acids show low solubility in aqueous solutions. In order to solubilize and ingest the released hydrophobic fatty acids, the blowflies must use a solubilizer. We applied native PAGE, Edman degradation, cDNA cloning, and RT-PCR to characterize a protein that accumulated in the oral disk of the black blowfly, Phormia regina. In situ hybridization was carried out to localize the expression at the cellular level. A fluorescence competitive binding assay was used to identify potential ligands of this protein. A protein newly identified from P. regina (PregOBP56a) belonged to the classic odorant-binding protein (OBP) family. This gene was expressed in a cluster of cells that was localized between pseudotracheae on the oral disk, which are not accessory cells of the taste peg chemosensory sensilla that normally synthesize OBPs. At pH 7 and pH 6, PregOBP56a bound palmitic, stearic, oleic, and linoleic acids, that are mainly found in chicken meat. The binding affinity of PregOBP56a decreased at pH 5. We propose that PregOBP56a is a protein that solubilizes fatty acids during feeding and subsequently helps to deliver the fatty acids to the midgut where it may help in the process of reproduction. As such, PregOBP56a is a potential molecular target for controlling the blowfly.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
Sigma-Aldrich
N-Phenyl-1-naphthylamin, reagent grade, 98%
Supelco
Stearinsäure, analytical standard
Supelco
Stearinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Stearinsäure, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland